Assay Details
| Gene Symbol: ACXA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259082.1 | NP_001246011.1 | 1 - 2 | 122 | 68 | |
| NM_001259083.1 | NP_001246012.1 | 1 - 2 | 122 | 68 | ||
| NM_001273494.1 | NP_001260423.1 | 1 - 2 | 122 | 68 | ||
| NM_001298954.1 | NP_001285883.1 | 1 - 2 | 122 | 68 | ||
| NM_138990.2 | NP_620475.2 | 1 - 2 | 122 | 68 | ||
More Information
Set Membership: |
|

ACXA
Genomic Map