Assay Details
| Gene Symbol: ACXA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259082.1 | NP_001246011.1 | 4 - 5 | 1207 | 78 | |
| NM_001273494.1 | NP_001260423.1 | 4 - 5 | 1207 | 78 | ||
| NM_138990.2 | NP_620475.2 | 4 - 5 | 1207 | 78 | ||
More Information
Set Membership: |
|

ACXA
Genomic Map