Assay Details
| Gene Symbol: ACXA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259082.1 | NP_001246011.1 | 9 - 10 | 2561 | 138 | |
| NM_001259083.1 | NP_001246012.1 | 9 - 10 | 2556 | 138 | ||
| NM_138990.2 | NP_620475.2 | 9 - 10 | 2561 | 138 | ||
More Information
Set Membership: |
|

ACXA
Genomic Map