Assay Details
| Gene Symbol: SLO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103788.3 | NP_001097258.2 | 2 | 630 | 63 | |
| NM_001103789.3 | NP_001097259.2 | 2 | 630 | 63 | ||
More Information
Set Membership: |
|

SLO2
Genomic Map