Assay Details
| Gene Symbol: COX7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299339.1 | NP_001286268.1 | 3 - 4 | 598 | 68 | |
| NM_001299340.1 | NP_001286269.1 | 2 - 3 | 375 | 68 | ||
| NM_144140.3 | NP_652397.1 | 1 - 2 | 168 | 68 | ||
| NM_176118.3 | NP_788298.1 | 1 - 2 | 168 | 68 | ||
| GenBank mRNA | AY118515.1 | - | 1 - 2 | 141 | 68 | |
More Information
Set Membership: |
|

COX7C
Genomic Map