Assay Details
| Gene Symbol: COX7C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299339.1 | NP_001286268.1 | 4 - 5 | 748 | 58 | |
| NM_144140.3 | NP_652397.1 | 2 - 3 | 318 | 58 | ||
| GenBank mRNA | AY118515.1 | - | 2 - 3 | 291 | 58 | |
More Information
Set Membership: |
|

COX7C
Genomic Map