Assay Details
| Gene Symbol: Nplp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300130.1 | NP_001287059.1 | 1 - 2 | 269 | 84 | |
| NM_001316458.1 | NP_001303387.1 | 1 - 2 | 111 | 84 | ||
| NM_144173.3 | NP_652430.1 | 1 - 2 | 111 | 84 | ||
| GenBank mRNA | AY113560.1 | - | 1 - 2 | 111 | 84 | |
| KX531956.1 | - | 1 - 2 | 46 | 84 | ||
More Information
Set Membership: |
|

Nplp2
Genomic Map