Assay Details
Gene Symbol: Neb-cGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272479.1 | NP_001259408.1 | 2 - 3 | 190 | 77 | |
NM_144291.4 | NP_652548.1 | 2 - 3 | 190 | 77 | ||
GenBank mRNA | AY071289.1 | - | 2 - 3 | 181 | 77 | |
AY071736.1 | - | 2 - 3 | 182 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
