Assay Details
| Gene Symbol: PIG-Q | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298394.1 | NP_001285323.1 | 4 - 5 | 1239 | 139 | |
| NM_144473.5 | NP_652730.4 | 4 - 5 | 1108 | 139 | ||
More Information
Set Membership: |
|

PIG-Q
Genomic Map