Assay Details
| Gene Symbol: PIG-Q | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298394.1 | NP_001285323.1 | 7 - 8 | 1531 | 114 | |
| NM_144473.5 | NP_652730.4 | 7 - 8 | 1400 | 114 | ||
More Information
Set Membership: |
|

PIG-Q
Genomic Map