Assay Details
Gene Symbol: AGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259385.1 | NP_001246314.1 | 4 - 5 | 542 | 69 | |
NM_079010.4 | NP_523734.1 | 3 - 4 | 414 | 69 | ||
NM_166020.2 | NP_725341.1 | 3 - 4 | 285 | 69 | ||
NM_166021.2 | NP_725342.1 | 4 - 5 | 542 | 69 | ||
GenBank mRNA | AB035447.1 | - | 3 - 4 | 385 | 69 | |
BT023945.1 | - | 3 - 4 | 285 | 69 | ||
BT100007.1 | - | 2 - 3 | 183 | 69 | ||
BT125895.1 | - | 4 - 5 | 542 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
