Assay Details
| Gene Symbol: CG8547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299486.1 | NP_001286415.1 | 6 - 7 | 1926 | 74 | |
| NM_137103.4 | NP_610947.3 | 6 - 7 | 1962 | 74 | ||
| NM_166044.3 | NP_725373.2 | 6 - 7 | 1900 | 74 | ||
More Information
Set Membership: |
|

CG8547
Genomic Map