Assay Details
Gene Symbol: Obp50d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166053.3 | NP_725388.2 | 1 - 2 | 104 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
