Assay Details
Gene Symbol: Strn-Mlck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202020.1 | NP_001188949.1 | 1 - 2 | 260 | 71 | |
NM_001202026.2 | NP_001188955.2 | 15 - 16 | 20475 | 71 | ||
NM_001274089.1 | NP_001261018.1 | 15 - 16 | 20475 | 71 | ||
NM_001274090.1 | NP_001261019.1 | 15 - 16 | 20843 | 71 | ||
NM_166129.3 | NP_725510.2 | 15 - 16 | 20843 | 71 | ||
GenBank mRNA | AY113646.1 | - | 1 - 2 | 261 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001202020.1 transcripts.
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
