Assay Details
Gene Symbol: Strn-Mlck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202023.2 | NP_001188952.1 | 4 - 5 | 1287 | 61 | |
NM_001202024.2 | NP_001188953.2 | 4 - 5 | 1322 | 61 | ||
NM_001202026.2 | NP_001188955.2 | 22 - 23 | 23298 | 61 | ||
NM_001274089.1 | NP_001261018.1 | 22 - 23 | 23298 | 61 | ||
NM_001274090.1 | NP_001261019.1 | 22 - 23 | 23666 | 61 | ||
NM_079030.4 | NP_523754.2 | 4 - 5 | 1276 | 61 | ||
NM_166128.3 | NP_725509.2 | 3 - 4 | 1337 | 61 | ||
NM_166129.3 | NP_725510.2 | 22 - 23 | 23666 | 61 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
