Assay Details
Gene Symbol: Strn-Mlck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202023.2 | NP_001188952.1 | 6 - 7 | 1727 | 98 | |
NM_001202024.2 | NP_001188953.2 | 6 - 7 | 1762 | 98 | ||
NM_001202026.2 | NP_001188955.2 | 24 - 25 | 23738 | 98 | ||
NM_001274089.1 | NP_001261018.1 | 24 - 25 | 23738 | 98 | ||
NM_001274090.1 | NP_001261019.1 | 24 - 25 | 24106 | 98 | ||
NM_079030.4 | NP_523754.2 | 6 - 7 | 1716 | 98 | ||
NM_166128.3 | NP_725509.2 | 5 - 6 | 1777 | 98 | ||
NM_166129.3 | NP_725510.2 | 24 - 25 | 24106 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
