Assay Details
Gene Symbol: Strn-Mlck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202023.2 | NP_001188952.1 | 7 - 8 | 1905 | 68 | |
NM_001274089.1 | NP_001261018.1 | 25 - 26 | 23916 | 68 | ||
NM_001274090.1 | NP_001261019.1 | 25 - 26 | 24284 | 68 | ||
NM_079030.4 | NP_523754.2 | 7 - 8 | 1894 | 68 | ||
NM_166128.3 | NP_725509.2 | 6 - 7 | 1955 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
