Assay Details
Gene Symbol: Strn-Mlck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202023.2 | NP_001188952.1 | 8 - 9 | 2619 | 54 | |
NM_001274089.1 | NP_001261018.1 | 26 - 27 | 24630 | 54 | ||
NM_001274090.1 | NP_001261019.1 | 26 - 27 | 24998 | 54 | ||
NM_079030.4 | NP_523754.2 | 8 - 9 | 2608 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
