Assay Details
Gene Symbol: Strn-Mlck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202026.2 | NP_001188955.2 | 5 - 6 | 1285 | 99 | |
NM_001274089.1 | NP_001261018.1 | 5 - 6 | 1285 | 99 | ||
NM_001274090.1 | NP_001261019.1 | 5 - 6 | 1653 | 99 | ||
NM_166129.3 | NP_725510.2 | 5 - 6 | 1653 | 99 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
