Assay Details
Gene Symbol: Strn-Mlck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166130.2 | NP_725511.2 | 3 - 4 | 3417 | 64 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
