Assay Details
Gene Symbol: Dg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079032.4 | NP_523756.1 | 3 - 4 | 955 | 76 | |
NM_166138.5 | NP_725523.3 | 3 - 4 | 955 | 76 | ||
NM_176183.3 | NP_788363.1 | 3 - 4 | 955 | 76 | ||
NM_176184.4 | NP_788364.2 | 3 - 4 | 955 | 76 | ||
GenBank mRNA | AF461100.1 | - | 2 - 3 | 307 | 76 | |
AF461101.1 | - | 2 - 3 | 307 | 76 | ||
AY069076.1 | - | 3 - 4 | 928 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
