Assay Details
| Gene Symbol: Syx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168905.2 | NP_730632.1 | 3 - 4 | 442 | 61 | |
| NM_168906.2 | NP_730633.1 | 3 - 4 | 479 | 61 | ||
| GenBank mRNA | AY069516.1 | - | 3 - 4 | 406 | 61 | |
More Information
Set Membership: |
|

Syx7
Genomic Map