Assay Details
| Gene Symbol: Syx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168905.2 | NP_730632.1 | 4 - 5 | 578 | 59 | |
| NM_168906.2 | NP_730633.1 | 4 - 5 | 615 | 59 | ||
| GenBank mRNA | AY069516.1 | - | 4 - 5 | 542 | 59 | |
More Information
Set Membership: |
|

Syx7
Genomic Map