Assay Details
Gene Symbol: EMC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168925.2 | NP_730661.2 | 2 - 3 | 313 | 72 | |
NM_168926.3 | NP_730662.2 | 2 - 3 | 298 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
