Assay Details
Gene Symbol: dlp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079347.3 | NP_524071.2 | 6 - 7 | 2150 | 63 | |
NM_206353.2 | NP_996075.2 | 6 - 7 | 2150 | 63 | ||
GenBank mRNA | AY052017.1 | - | 6 - 7 | 2131 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
