Assay Details
Gene Symbol: eIF2Bepsilon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130605.5 | NP_569961.2 | 1 - 2 | 149 | 83 | |
NM_206606.3 | NP_996329.1 | 1 - 2 | 149 | 83 | ||
GenBank mRNA | AY095053.1 | - | 1 - 2 | 100 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
