Assay Details
Gene Symbol: eIF2Bepsilon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130605.5 | NP_569961.2 | 3 - 4 | 1553 | 76 | |
NM_206606.3 | NP_996329.1 | 3 - 4 | 1553 | 76 | ||
GenBank mRNA | AJ344150.1 | - | 3 - 4 | 1525 | 76 | |
AY095053.1 | - | 3 - 4 | 1504 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
