Assay Details
Gene Symbol: sgg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042794.4 | NP_001036259.2 | 3 - 4 | 1292 | 134 | |
NM_001169178.1 | NP_001162649.1 | 4 - 5 | 917 | 134 | ||
NM_001169179.1 | NP_001162650.1 | 3 - 4 | 539 | 134 | ||
NM_001272263.1 | NP_001259192.1 | 4 - 5 | 2363 | 134 | ||
NM_001272264.1 | NP_001259193.1 | 4 - 5 | 2494 | 134 | ||
NM_057366.5 | NP_476714.1 | 3 - 4 | 924 | 134 | ||
NM_057367.5 | NP_476715.1 | 3 - 4 | 563 | 134 | ||
NM_057368.5 | NP_476716.2 | 2 - 3 | 2521 | 134 | ||
NM_134278.3 | NP_599105.1 | 3 - 4 | 826 | 134 | ||
NM_166947.3 | NP_726822.1 | 3 - 4 | 771 | 134 | ||
NM_166948.4 | NP_726823.1 | 3 - 4 | 924 | 134 | ||
NM_206611.2 | NP_996334.1 | 2 - 3 | 480 | 134 | ||
NM_206612.3 | NP_996335.1 | 2 - 3 | 909 | 134 | ||
NM_206613.3 | NP_996336.1 | 5 - 6 | 1213 | 134 | ||
NM_206614.3 | NP_996337.1 | 4 - 5 | 807 | 134 | ||
NM_206615.2 | NP_996338.1 | 3 - 4 | 737 | 134 | ||
GenBank mRNA | AY122193.1 | - | 3 - 4 | 899 | 134 | |
BT050474.1 | - | 2 - 3 | 859 | 134 | ||
BT072831.1 | - | 3 - 4 | 548 | 134 | ||
BT133153.1 | - | 2 - 3 | 2515 | 134 | ||
X70863.1 | - | 883 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT072831.1, NM_001169178.1, NM_001169179.1, NM_206611.2 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
