Assay Details
Gene Symbol: sgg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042794.4 | NP_001036259.2 | 4 - 5 | 1372 | 126 | |
NM_001169178.1 | NP_001162649.1 | 5 - 6 | 997 | 126 | ||
NM_001169179.1 | NP_001162650.1 | 4 - 5 | 619 | 126 | ||
NM_001272263.1 | NP_001259192.1 | 5 - 6 | 2443 | 126 | ||
NM_001272264.1 | NP_001259193.1 | 5 - 6 | 2574 | 126 | ||
NM_057366.5 | NP_476714.1 | 4 - 5 | 1004 | 126 | ||
NM_057367.5 | NP_476715.1 | 4 - 5 | 643 | 126 | ||
NM_057368.5 | NP_476716.2 | 3 - 4 | 2601 | 126 | ||
NM_134278.3 | NP_599105.1 | 4 - 5 | 906 | 126 | ||
NM_166947.3 | NP_726822.1 | 4 - 5 | 851 | 126 | ||
NM_166948.4 | NP_726823.1 | 4 - 5 | 1004 | 126 | ||
NM_206611.2 | NP_996334.1 | 3 - 4 | 560 | 126 | ||
NM_206612.3 | NP_996335.1 | 3 - 4 | 989 | 126 | ||
NM_206613.3 | NP_996336.1 | 6 - 7 | 1293 | 126 | ||
NM_206614.3 | NP_996337.1 | 5 - 6 | 887 | 126 | ||
NM_206615.2 | NP_996338.1 | 4 - 5 | 817 | 126 | ||
GenBank mRNA | AY122193.1 | - | 4 - 5 | 979 | 126 | |
BT050474.1 | - | 3 - 4 | 939 | 126 | ||
BT072831.1 | - | 4 - 5 | 628 | 126 | ||
BT133153.1 | - | 3 - 4 | 2595 | 126 | ||
X70863.1 | - | 963 | 126 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
