Assay Details
Gene Symbol: sgg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042794.4 | NP_001036259.2 | 6 - 7 | 1725 | 75 | |
NM_001103400.2 | NP_001096870.1 | 6 - 7 | 1331 | 75 | ||
NM_001169178.1 | NP_001162649.1 | 7 - 8 | 1350 | 75 | ||
NM_001169179.1 | NP_001162650.1 | 6 - 7 | 972 | 75 | ||
NM_001272263.1 | NP_001259192.1 | 7 - 8 | 2796 | 75 | ||
NM_001272264.1 | NP_001259193.1 | 7 - 8 | 2927 | 75 | ||
NM_057366.5 | NP_476714.1 | 6 - 7 | 1357 | 75 | ||
NM_057367.5 | NP_476715.1 | 6 - 7 | 996 | 75 | ||
NM_057368.5 | NP_476716.2 | 5 - 6 | 2954 | 75 | ||
NM_134278.3 | NP_599105.1 | 6 - 7 | 1259 | 75 | ||
NM_166947.3 | NP_726822.1 | 6 - 7 | 1204 | 75 | ||
NM_166948.4 | NP_726823.1 | 6 - 7 | 1357 | 75 | ||
NM_206611.2 | NP_996334.1 | 5 - 6 | 913 | 75 | ||
NM_206612.3 | NP_996335.1 | 5 - 6 | 1342 | 75 | ||
NM_206613.3 | NP_996336.1 | 8 - 9 | 1646 | 75 | ||
NM_206614.3 | NP_996337.1 | 7 - 8 | 1240 | 75 | ||
NM_206615.2 | NP_996338.1 | 6 - 7 | 1170 | 75 | ||
GenBank mRNA | AY122193.1 | - | 6 - 7 | 1332 | 75 | |
BT050474.1 | - | 5 - 6 | 1292 | 75 | ||
BT072831.1 | - | 6 - 7 | 981 | 75 | ||
BT133153.1 | - | 5 - 6 | 2948 | 75 | ||
X70863.1 | - | 1316 | 75 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
