Assay Details
Gene Symbol: sgg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042794.4 | NP_001036259.2 | 7 - 8 | 1915 | 110 | |
NM_001103400.2 | NP_001096870.1 | 7 - 8 | 1521 | 110 | ||
NM_001169178.1 | NP_001162649.1 | 8 - 9 | 1540 | 110 | ||
NM_001169179.1 | NP_001162650.1 | 7 - 8 | 1162 | 110 | ||
NM_001272264.1 | NP_001259193.1 | 8 - 9 | 3117 | 110 | ||
NM_057366.5 | NP_476714.1 | 7 - 8 | 1547 | 110 | ||
NM_057367.5 | NP_476715.1 | 7 - 8 | 1186 | 110 | ||
NM_057368.5 | NP_476716.2 | 6 - 7 | 3144 | 110 | ||
NM_134278.3 | NP_599105.1 | 7 - 8 | 1449 | 110 | ||
NM_166947.3 | NP_726822.1 | 7 - 8 | 1394 | 110 | ||
NM_166948.4 | NP_726823.1 | 7 - 8 | 1547 | 110 | ||
NM_206611.2 | NP_996334.1 | 6 - 7 | 1103 | 110 | ||
NM_206612.3 | NP_996335.1 | 6 - 7 | 1532 | 110 | ||
NM_206613.3 | NP_996336.1 | 9 - 10 | 1836 | 110 | ||
NM_206614.3 | NP_996337.1 | 8 - 9 | 1430 | 110 | ||
NM_206615.2 | NP_996338.1 | 7 - 8 | 1360 | 110 | ||
GenBank mRNA | AY122193.1 | - | 7 - 8 | 1522 | 110 | |
BT050474.1 | - | 6 - 7 | 1482 | 110 | ||
BT072831.1 | - | 7 - 8 | 1171 | 110 | ||
BT133153.1 | - | 6 - 7 | 3138 | 110 | ||
X70863.1 | - | 1506 | 110 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
