Assay Details
Gene Symbol: sgg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057366.5 | NP_476714.1 | 2 - 3 | 734 | 86 | |
NM_057367.5 | NP_476715.1 | 2 - 3 | 373 | 86 | ||
NM_134278.3 | NP_599105.1 | 2 - 3 | 636 | 86 | ||
NM_166947.3 | NP_726822.1 | 2 - 3 | 581 | 86 | ||
NM_166948.4 | NP_726823.1 | 2 - 3 | 734 | 86 | ||
NM_206613.3 | NP_996336.1 | 4 - 5 | 1023 | 86 | ||
NM_206614.3 | NP_996337.1 | 3 - 4 | 617 | 86 | ||
NM_206615.2 | NP_996338.1 | 2 - 3 | 547 | 86 | ||
GenBank mRNA | AY122193.1 | - | 2 - 3 | 709 | 86 | |
X70863.1 | - | 693 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
