Assay Details
Gene Symbol: Vap33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130731.3 | NP_570087.1 | 3 - 4 | 791 | 71 | |
NM_206623.2 | NP_996346.2 | 3 - 4 | 791 | 71 | ||
NM_206624.2 | NP_996347.1 | 3 - 4 | 501 | 71 | ||
NM_206625.2 | NP_996348.1 | 3 - 4 | 791 | 71 | ||
GenBank mRNA | AY060395.1 | - | 3 - 4 | 762 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
