Assay Details
| Gene Symbol: sdt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038746.3 | NP_001033835.2 | 11 - 12 | 5254 | 87 | |
| NM_001038747.3 | NP_001033836.1 | 6 - 7 | 1274 | 87 | ||
| NM_001042798.2 | NP_001036263.1 | 4 - 5 | 809 | 87 | ||
| NM_001258645.1 | NP_001245574.1 | 11 - 12 | 3141 | 87 | ||
| NM_001258647.1 | NP_001245576.1 | 12 - 13 | 4480 | 87 | ||
| NM_001272397.1 | NP_001259326.1 | 11 - 12 | 4897 | 87 | ||
| NM_001272398.1 | NP_001259327.1 | 6 - 7 | 2442 | 87 | ||
| NM_001298081.1 | NP_001285010.1 | 10 - 11 | 4012 | 87 | ||
| NM_001298082.1 | NP_001285011.1 | 9 - 10 | 2713 | 87 | ||
| NM_132235.5 | NP_572463.1 | 7 - 8 | 2573 | 87 | ||
| NM_132236.6 | NP_572464.4 | 6 - 7 | 2006 | 87 | ||
| NM_206653.3 | NP_996376.1 | 6 - 7 | 2235 | 87 | ||
| GenBank mRNA | BT010281.1 | - | 6 - 7 | 1272 | 87 | |
More Information
Set Membership: |
|

sdt
Genomic Map