Assay Details
Gene Symbol: sdt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038746.3 | NP_001033835.2 | 15 - 16 | 7107 | 67 | |
NM_001038747.3 | NP_001033836.1 | 10 - 11 | 3127 | 67 | ||
NM_001042798.2 | NP_001036263.1 | 8 - 9 | 2662 | 67 | ||
NM_001258645.1 | NP_001245574.1 | 16 - 17 | 5051 | 67 | ||
NM_001258646.1 | NP_001245575.1 | 6 - 7 | 2435 | 67 | ||
NM_001258647.1 | NP_001245576.1 | 17 - 18 | 6390 | 67 | ||
NM_001272397.1 | NP_001259326.1 | 15 - 16 | 6750 | 67 | ||
NM_001272398.1 | NP_001259327.1 | 10 - 11 | 4295 | 67 | ||
NM_001298081.1 | NP_001285010.1 | 14 - 15 | 5865 | 67 | ||
NM_001298082.1 | NP_001285011.1 | 13 - 14 | 4566 | 67 | ||
NM_132235.5 | NP_572463.1 | 11 - 12 | 4426 | 67 | ||
NM_132236.6 | NP_572464.4 | 10 - 11 | 3859 | 67 | ||
NM_206652.2 | NP_996375.1 | 6 - 7 | 2483 | 67 | ||
NM_206653.3 | NP_996376.1 | 10 - 11 | 4088 | 67 | ||
GenBank mRNA | BT010281.1 | - | 10 - 11 | 3125 | 67 | |
BT099747.1 | - | 6 - 7 | 2483 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
