Assay Details
| Gene Symbol: sdt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038746.3 | NP_001033835.2 | 2 - 3 | 1505 | 65 | |
| NM_001258645.1 | NP_001245574.1 | 3 - 4 | 928 | 65 | ||
| NM_001258647.1 | NP_001245576.1 | 3 - 4 | 1025 | 65 | ||
| NM_001272397.1 | NP_001259326.1 | 2 - 3 | 1505 | 65 | ||
| NM_001298081.1 | NP_001285010.1 | 2 - 3 | 1505 | 65 | ||
| NM_001298082.1 | NP_001285011.1 | 2 - 3 | 1505 | 65 | ||
More Information
Set Membership: |
|

sdt
Genomic Map