Assay Details
| Gene Symbol: sdt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038746.3 | NP_001033835.2 | 9 - 10 | 3727 | 104 | |
| NM_001272397.1 | NP_001259326.1 | 9 - 10 | 3727 | 104 | ||
| NM_001272398.1 | NP_001259327.1 | 4 - 5 | 915 | 104 | ||
| NM_001298081.1 | NP_001285010.1 | 8 - 9 | 2485 | 104 | ||
| NM_132235.5 | NP_572463.1 | 5 - 6 | 1046 | 104 | ||
| NM_206653.3 | NP_996376.1 | 4 - 5 | 708 | 104 | ||
More Information
Set Membership: |
|

sdt
Genomic Map