Assay Details
Gene Symbol: Trf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103435.3 | NP_001096905.1 | 7 - 8 | 5015 | 73 | |
NM_001169230.2 | NP_001162701.1 | 6 - 7 | 4934 | 73 | ||
NM_001272400.1 | NP_001259329.1 | 7 - 8 | 4984 | 73 | ||
NM_001272401.1 | NP_001259330.1 | 5 - 6 | 4724 | 73 | ||
NM_078529.4 | NP_511084.2 | 5 - 6 | 4818 | 73 | ||
NM_206654.2 | NP_996377.1 | 7 - 8 | 5109 | 73 | ||
GenBank mRNA | AY069554.1 | - | 2 - 3 | 2389 | 73 | |
DQ162845.1 | - | 7 - 8 | 4995 | 73 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
