Assay Details
Gene Symbol: AP-1gamma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103448.3 | NP_001096918.1 | 9 - 10 | 2189 | 114 | |
NM_001258660.1 | NP_001245589.1 | 8 - 9 | 1931 | 114 | ||
NM_001258661.2 | NP_001245590.1 | 10 - 11 | 2207 | 114 | ||
NM_001272442.1 | NP_001259371.1 | 9 - 10 | 2100 | 114 | ||
NM_132299.6 | NP_572527.2 | 9 - 10 | 1958 | 114 | ||
NM_167178.5 | NP_727311.2 | 8 - 9 | 1863 | 114 | ||
NM_176717.4 | NP_788890.2 | 10 - 11 | 1976 | 114 | ||
NM_176718.4 | NP_788891.2 | 9 - 10 | 1881 | 114 | ||
NM_206671.4 | NP_996394.2 | 9 - 10 | 1896 | 114 | ||
GenBank mRNA | BT100000.1 | - | 10 - 11 | 2202 | 114 | |
BT100006.1 | - | 8 - 9 | 1932 | 114 | ||
BT100128.1 | - | 9 - 10 | 1944 | 114 | ||
BT133052.1 | - | 8 - 9 | 1861 | 114 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
