Assay Details
| Gene Symbol: AP-1gamma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103448.3 | NP_001096918.1 | 11 - 12 | 2609 | 99 | |
| NM_001258660.1 | NP_001245589.1 | 10 - 11 | 2351 | 99 | ||
| NM_001258661.2 | NP_001245590.1 | 12 - 13 | 2627 | 99 | ||
| NM_001272442.1 | NP_001259371.1 | 11 - 12 | 2520 | 99 | ||
| NM_132299.6 | NP_572527.2 | 11 - 12 | 2378 | 99 | ||
| NM_167178.5 | NP_727311.2 | 10 - 11 | 2283 | 99 | ||
| NM_176717.4 | NP_788890.2 | 12 - 13 | 2396 | 99 | ||
| NM_176718.4 | NP_788891.2 | 11 - 12 | 2301 | 99 | ||
| NM_206671.4 | NP_996394.2 | 11 - 12 | 2316 | 99 | ||
| GenBank mRNA | BT100000.1 | - | 12 - 13 | 2622 | 99 | |
| BT100128.1 | - | 11 - 12 | 2364 | 99 | ||
| BT133052.1 | - | 10 - 11 | 2281 | 99 | ||
More Information
Set Membership: |
|

AP-1gamma
Genomic Map