Assay Details
Gene Symbol: AP-1gamma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103448.3 | NP_001096918.1 | 14 - 15 | 3384 | 80 | |
NM_001258660.1 | NP_001245589.1 | 13 - 14 | 3126 | 80 | ||
NM_001258661.2 | NP_001245590.1 | 15 - 16 | 3402 | 80 | ||
NM_001272442.1 | NP_001259371.1 | 14 - 15 | 3295 | 80 | ||
NM_132299.6 | NP_572527.2 | 14 - 15 | 3153 | 80 | ||
NM_167178.5 | NP_727311.2 | 13 - 14 | 3058 | 80 | ||
NM_176717.4 | NP_788890.2 | 15 - 16 | 3171 | 80 | ||
NM_176718.4 | NP_788891.2 | 14 - 15 | 3076 | 80 | ||
NM_206671.4 | NP_996394.2 | 14 - 15 | 3091 | 80 | ||
GenBank mRNA | BT100000.1 | - | 15 - 16 | 3397 | 80 | |
BT100006.1 | - | 13 - 14 | 3127 | 80 | ||
BT100128.1 | - | 14 - 15 | 3139 | 80 | ||
BT133052.1 | - | 13 - 14 | 3056 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
