Assay Details
Gene Symbol: AP-1gamma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258661.2 | NP_001245590.1 | 7 - 8 | 1333 | 61 | |
NM_176717.4 | NP_788890.2 | 7 - 8 | 1102 | 61 | ||
NM_176718.4 | NP_788891.2 | 6 - 7 | 1007 | 61 | ||
GenBank mRNA | BT100000.1 | - | 7 - 8 | 1328 | 61 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
