Assay Details
Gene Symbol: alpha-Man-Ia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272473.2 | NP_001259402.1 | 3 - 4 | 1448 | 88 | |
NM_078550.4 | NP_511105.2 | 2 - 3 | 1273 | 88 | ||
NM_167223.3 | NP_727407.1 | 3 - 4 | 1695 | 88 | ||
NM_167224.3 | NP_727408.2 | 2 - 3 | 1831 | 88 | ||
NM_206672.3 | NP_996395.1 | 3 - 4 | 1737 | 88 | ||
NM_206673.3 | NP_996396.1 | 3 - 4 | 1537 | 88 | ||
NM_206674.3 | NP_996397.1 | 3 - 4 | 1582 | 88 | ||
NM_206675.3 | NP_996398.1 | 3 - 4 | 1690 | 88 | ||
NM_206676.3 | NP_996399.1 | 3 - 4 | 1514 | 88 | ||
GenBank mRNA | BT011036.1 | - | 3 - 4 | 1694 | 88 | |
BT024451.1 | - | 2 - 3 | 1831 | 88 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
