Assay Details
Gene Symbol: alpha-Man-Ia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272473.2 | NP_001259402.1 | 5 - 6 | 1804 | 102 | |
NM_078550.4 | NP_511105.2 | 4 - 5 | 1629 | 102 | ||
NM_167223.3 | NP_727407.1 | 5 - 6 | 2051 | 102 | ||
NM_167224.3 | NP_727408.2 | 4 - 5 | 2187 | 102 | ||
NM_206672.3 | NP_996395.1 | 5 - 6 | 2093 | 102 | ||
NM_206673.3 | NP_996396.1 | 5 - 6 | 1893 | 102 | ||
NM_206674.3 | NP_996397.1 | 5 - 6 | 1938 | 102 | ||
NM_206675.3 | NP_996398.1 | 5 - 6 | 2046 | 102 | ||
NM_206676.3 | NP_996399.1 | 5 - 6 | 1870 | 102 | ||
GenBank mRNA | BT024451.1 | - | 4 - 5 | 2187 | 102 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
