Assay Details
Gene Symbol: CkIIbeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014731.2 | NP_001014731.1 | 3 - 4 | 292 | 67 | |
NM_001258699.1 | NP_001245628.1 | 3 - 4 | 211 | 67 | ||
NM_001258700.2 | NP_001245629.1 | 3 - 4 | 580 | 67 | ||
NM_001258701.2 | NP_001245630.1 | 3 - 4 | 314 | 67 | ||
NM_001272533.1 | NP_001259462.1 | 3 - 4 | 580 | 67 | ||
NM_078576.5 | NP_511131.3 | 3 - 4 | 246 | 67 | ||
NM_080762.4 | NP_542940.1 | 3 - 4 | 333 | 67 | ||
NM_167304.3 | NP_727561.1 | 3 - 4 | 580 | 67 | ||
NM_167305.3 | NP_727562.1 | 3 - 4 | 215 | 67 | ||
NM_206692.3 | NP_996415.1 | 3 - 4 | 333 | 67 | ||
GenBank mRNA | AY113447.1 | - | 3 - 4 | 330 | 67 | |
BT120031.1 | - | 3 - 4 | 549 | 67 | ||
M16535.1 | - | 3 - 4 | 197 | 67 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
