Assay Details
| Gene Symbol: CkIIbeta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014731.2 | NP_001014731.1 | 5 - 6 | 671 | 87 | |
| NM_001258699.1 | NP_001245628.1 | 5 - 6 | 590 | 87 | ||
| NM_001258700.2 | NP_001245629.1 | 5 - 6 | 959 | 87 | ||
| NM_001258701.2 | NP_001245630.1 | 5 - 6 | 693 | 87 | ||
| NM_001272533.1 | NP_001259462.1 | 5 - 6 | 959 | 87 | ||
| NM_078576.5 | NP_511131.3 | 5 - 6 | 625 | 87 | ||
| NM_080762.4 | NP_542940.1 | 5 - 6 | 712 | 87 | ||
| NM_167304.3 | NP_727561.1 | 5 - 6 | 959 | 87 | ||
| NM_167305.3 | NP_727562.1 | 5 - 6 | 594 | 87 | ||
| NM_206692.3 | NP_996415.1 | 5 - 6 | 712 | 87 | ||
| GenBank mRNA | AY113447.1 | - | 5 - 6 | 709 | 87 | |
| BT120031.1 | - | 5 - 6 | 928 | 87 | ||
| M16535.1 | - | 5 - 6 | 576 | 87 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  3' Most  Drosophila melanogaster | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 CkIIbeta
CkIIbeta Genomic Map
Genomic Map Back To Top
 Back To Top