Assay Details
Gene Symbol: Chc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103523.2 | NP_001096993.1 | 2 - 3 | 400 | 57 | |
NM_001103524.2 | NP_001096994.1 | 2 - 3 | 294 | 57 | ||
NM_001298371.1 | NP_001285300.1 | 3 - 4 | 505 | 57 | ||
NM_057694.3 | NP_477042.1 | 2 - 3 | 413 | 57 | ||
NM_167466.2 | NP_727901.1 | 2 - 3 | 370 | 57 | ||
NM_206728.2 | NP_996451.1 | 2 - 3 | 510 | 57 | ||
NM_206729.2 | NP_996452.1 | 2 - 3 | 451 | 57 | ||
GenBank mRNA | AY119615.1 | - | 2 - 3 | 397 | 57 | |
Z14133.1 | - | 327 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
