Assay Details
Gene Symbol: Chc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103523.2 | NP_001096993.1 | 6 - 7 | 5166 | 66 | |
NM_001103524.2 | NP_001096994.1 | 6 - 7 | 5060 | 66 | ||
NM_001298371.1 | NP_001285300.1 | 7 - 8 | 5271 | 66 | ||
NM_057694.3 | NP_477042.1 | 6 - 7 | 5179 | 66 | ||
NM_167466.2 | NP_727901.1 | 6 - 7 | 5136 | 66 | ||
NM_206728.2 | NP_996451.1 | 6 - 7 | 5276 | 66 | ||
NM_206729.2 | NP_996452.1 | 6 - 7 | 5217 | 66 | ||
GenBank mRNA | AY119615.1 | - | 6 - 7 | 5163 | 66 | |
Z14133.1 | - | 5093 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
