Assay Details
Gene Symbol: Myb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078615.4 | NP_511170.1 | 2 - 3 | 1934 | 68 | |
NM_206731.2 | NP_996454.1 | 1 - 2 | 2029 | 68 | ||
NM_206733.2 | NP_996456.1 | 2 - 3 | 1966 | 68 | ||
NM_206734.2 | NP_996457.1 | 2 - 3 | 1858 | 68 | ||
GenBank mRNA | BT003262.1 | - | 1 - 2 | 1977 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
