Assay Details
Gene Symbol: Stim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258774.2 | NP_001245703.1 | 7 - 8 | 1877 | 60 | |
NM_078633.4 | NP_523357.2 | 7 - 8 | 1813 | 60 | ||
NM_206747.3 | NP_996470.2 | 7 - 8 | 1813 | 60 | ||
GenBank mRNA | AF328906.1 | - | 6 - 7 | 1769 | 60 | |
AY069686.1 | - | 7 - 8 | 1797 | 60 | ||
BT133089.1 | - | 7 - 8 | 1791 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
